Assay Details
Gene Symbol: PHI-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103210.4 | NP_174746.1 | 1 | 946 | 65 | |
GenBank mRNA | AY084831.1 | - | 1 | 933 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
