Assay Details
| Gene Symbol: AT1G35150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103212.1 | NP_174748.1 | 3 - 4 | 1120 | 122 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G35150| Gene Symbol: AT1G35150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103212.1 | NP_174748.1 | 3 - 4 | 1120 | 122 | |
Set Membership: |
|