Assay Details
Gene Symbol: ACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198214.2 | NP_001185143.1 | 24 - 25 | 3401 | 120 | |
NM_103313.4 | NP_174849.2 | 24 - 25 | 3372 | 120 | ||
GenBank mRNA | D34630.2 | - | 23 - 24 | 3011 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
