Assay Details
Gene Symbol: ACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198214.2 | NP_001185143.1 | 29 - 30 | 4796 | 109 | |
NM_103313.4 | NP_174849.2 | 29 - 30 | 4767 | 109 | ||
GenBank mRNA | D34630.2 | - | 28 - 29 | 4406 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
