Assay Details
Gene Symbol: ACC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198214.2 | NP_001185143.1 | 3 - 4 | 966 | 79 | |
NM_103313.4 | NP_174849.2 | 3 - 4 | 937 | 79 | ||
GenBank mRNA | D34630.2 | - | 2 - 3 | 576 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
