Assay Details
Gene Symbol: CH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333230.1 | NP_001321683.1 | 2 - 3 | 418 | 87 | |
NM_103548.5 | NP_175088.1 | 3 - 4 | 551 | 87 | ||
NM_202240.2 | NP_973969.1 | 3 - 4 | 504 | 87 | ||
NM_202241.2 | NP_973970.1 | 3 - 4 | 504 | 87 | ||
GenBank mRNA | AK228645.1 | - | 3 - 4 | 469 | 87 | |
AY128357.1 | - | 3 - 4 | 504 | 87 | ||
BT002075.1 | - | 3 - 4 | 432 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
