Assay Details
Gene Symbol: BGAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333243.1 | NP_001320473.1 | 6 - 7 | 696 | 70 | |
NM_103587.3 | NP_175127.1 | 8 - 9 | 1228 | 70 | ||
GenBank mRNA | AY069911.1 | - | 8 - 9 | 1027 | 70 | |
AY093977.1 | - | 8 - 9 | 892 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
