Assay Details
Gene Symbol: BGAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333243.1 | NP_001320473.1 | 7 - 8 | 780 | 104 | |
NM_103587.3 | NP_175127.1 | 9 - 10 | 1312 | 104 | ||
GenBank mRNA | AY069911.1 | - | 9 - 10 | 1111 | 104 | |
AY093977.1 | - | 9 - 10 | 976 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
