Assay Details
Gene Symbol: GAMMA CA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103620.4 | NP_175159.1 | 4 - 5 | 677 | 130 | |
GenBank mRNA | AY045979.1 | - | 4 - 5 | 646 | 130 | |
AY113936.1 | - | 4 - 5 | 540 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
