Assay Details
| Gene Symbol: AT1G48100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103706.4 | NP_175244.1 | 2 - 3 | 771 | 67 | |
| GenBank mRNA | AY050798.1 | - | 2 - 3 | 636 | 67 | |
| AY096360.1 | - | 2 - 3 | 588 | 67 | ||
More Information
Set Membership: |
|

AT1G48100
Genomic Map