Assay Details
| Gene Symbol: AT1G48100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_103706.4 | NP_175244.1 | 3 - 4 | 812 | 142 | |
| GenBank mRNA | AY050798.1 | - | 3 - 4 | 677 | 142 | |
| AY096360.1 | - | 3 - 4 | 629 | 142 | ||
More Information
Set Membership: |
|

AT1G48100
Genomic Map