Assay Details
| Gene Symbol: CIPK17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333316.1 | NP_001320279.1 | 8 - 9 | 2165 | 89 | |
| NM_001333317.1 | NP_001320280.1 | 7 - 8 | 1396 | 89 | ||
| NM_001333318.1 | NP_001320277.1 | 9 - 10 | 2007 | 89 | ||
| NM_001333319.1 | NP_001320278.1 | 9 - 10 | 1725 | 89 | ||
| NM_103723.4 | NP_175260.1 | 9 - 10 | 1279 | 89 | ||
| GenBank mRNA | AY036958.1 | - | 9 - 10 | 1099 | 89 | |
| BT029240.1 | - | 9 - 10 | 1099 | 89 | ||
More Information
Set Membership: |
|

CIPK17
Genomic Map