Assay Details
Gene Symbol: CHR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001333322.1 | NP_001321270.1 | 21 - 22 | 1943 | 83 | |
NM_001333323.1 | NP_001321271.1 | 21 - 22 | 1820 | 83 | ||
NM_103728.5 | NP_175265.3 | 21 - 22 | 1928 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
