Assay Details
| Gene Symbol: CHR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333322.1 | NP_001321270.1 | 23 - 24 | 2046 | 104 | |
| NM_103728.5 | NP_175265.3 | 23 - 24 | 2031 | 104 | ||
More Information
Set Membership: |
|

CHR18
Genomic Map