Assay Details
| Gene Symbol: CHR18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001333322.1 | NP_001321270.1 | 7 - 8 | 742 | 121 | |
| NM_001333323.1 | NP_001321271.1 | 7 - 8 | 619 | 121 | ||
| NM_001333324.1 | NP_001321269.1 | 7 - 8 | 721 | 121 | ||
| NM_103728.5 | NP_175265.3 | 7 - 8 | 727 | 121 | ||
More Information
Set Membership: |
|

CHR18
Genomic Map