Assay Details
| Gene Symbol: RBL15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203197.1 | NP_001190126.1 | 11 - 12 | 1200 | 76 | |
| NM_115708.4 | NP_567064.1 | 10 - 11 | 1333 | 76 | ||
| GenBank mRNA | AY136386.1 | - | 10 - 11 | 1167 | 76 | |
| BT000186.1 | - | 10 - 11 | 1072 | 76 | ||
More Information
Set Membership: |
|

RBL15
Genomic Map