Assay Details
Gene Symbol: PRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115784.3 | NP_191481.1 | 3 - 4 | 582 | 75 | |
GenBank mRNA | AF353716.1 | - | 3 - 4 | 260 | 75 | |
BT026056.1 | - | 3 - 4 | 189 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
