Assay Details
| Gene Symbol: GUN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115802.4 | NP_191499.1 | 1 | 848 | 63 | |
| GenBank mRNA | AY150510.1 | - | 1 | 730 | 63 | |
| Gene Symbol: GCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001339969.1 | NP_001326041.1 | 1 | 99 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001339969.1 transcripts.
Set Membership: |
|

GUN4
Genomic Map