Assay Details
| Gene Symbol: GCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203206.1 | NP_001190135.1 | 8 - 9 | 1546 | 144 | |
| NM_001339969.1 | NP_001326041.1 | 8 - 9 | 2413 | 144 | ||
| NM_001339970.1 | NP_001326042.1 | 7 - 8 | 1555 | 144 | ||
| NM_001339971.1 | NP_001326040.1 | 7 - 8 | 1326 | 144 | ||
| NM_115803.3 | NP_191500.2 | 7 - 8 | 1445 | 144 | ||
| GenBank mRNA | AJ459823.1 | - | 7 - 8 | 1093 | 144 | |
More Information
Set Membership: |
|

GCN2
Genomic Map