Assay Details
| Gene Symbol: GCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203206.1 | NP_001190135.1 | 10 - 11 | 1753 | 73 | |
| NM_001339969.1 | NP_001326041.1 | 10 - 11 | 2620 | 73 | ||
| NM_001339970.1 | NP_001326042.1 | 9 - 10 | 1762 | 73 | ||
| NM_001339971.1 | NP_001326040.1 | 9 - 10 | 1533 | 73 | ||
| NM_115803.3 | NP_191500.2 | 9 - 10 | 1652 | 73 | ||
| GenBank mRNA | AJ459823.1 | - | 9 - 10 | 1300 | 73 | |
More Information
Set Membership: |
|

GCN2
Genomic Map