Assay Details
Gene Symbol: SK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115865.2 | NP_567091.1 | 1 | 206 | 79 | |
GenBank mRNA | DQ056631.1 | - | 1 | 135 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
