Assay Details
| Gene Symbol: BGLU16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035822.2 | NP_001030899.1 | 6 - 7 | 626 | 86 | |
| NM_001084847.1 | NP_001078316.1 | 6 - 7 | 814 | 86 | ||
| NM_115876.5 | NP_191572.1 | 7 - 8 | 1093 | 86 | ||
| GenBank mRNA | AY045953.1 | - | 7 - 8 | 817 | 86 | |
| AY113935.1 | - | 7 - 8 | 776 | 86 | ||
More Information
Set Membership: |
|

BGLU16
Genomic Map