Assay Details
| Gene Symbol: DIN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001340024.1 | NP_001327679.1 | 7 - 8 | 1092 | 79 | |
| NM_115877.4 | NP_191573.1 | 7 - 8 | 1092 | 79 | ||
More Information
Set Membership: |
|

DIN2
Genomic Map