Assay Details
Gene Symbol: DL1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115882.4 | NP_567094.1 | 10 - 11 | 1333 | 59 | |
GenBank mRNA | AF428332.1 | - | 10 - 11 | 1155 | 59 | |
AJ304842.1 | - | 10 - 11 | 1115 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
