Assay Details
Gene Symbol: DL1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115882.4 | NP_567094.1 | 7 - 8 | 949 | 98 | |
GenBank mRNA | AF428332.1 | - | 7 - 8 | 771 | 98 | |
AJ304842.1 | - | 7 - 8 | 731 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
