Assay Details
Gene Symbol: DL1E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115882.4 | NP_567094.1 | 9 - 10 | 1215 | 140 | |
GenBank mRNA | AF428332.1 | - | 9 - 10 | 1037 | 140 | |
AJ304842.1 | - | 9 - 10 | 997 | 140 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
