Assay Details
Gene Symbol: EDA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115900.4 | NP_191595.1 | 4 - 5 | 463 | 125 | |
GenBank mRNA | AY039876.1 | - | 4 - 5 | 435 | 125 | |
AY085156.1 | - | 4 - 5 | 418 | 125 | ||
AY101514.1 | - | 3 - 4 | 244 | 125 | ||
BT000725.1 | - | 4 - 5 | 379 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
