Assay Details
Gene Symbol: EDA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115900.4 | NP_191595.1 | 5 - 6 | 552 | 92 | |
GenBank mRNA | AY039876.1 | - | 5 - 6 | 524 | 92 | |
AY085156.1 | - | 5 - 6 | 507 | 92 | ||
AY101514.1 | - | 4 - 5 | 333 | 92 | ||
BT000725.1 | - | 5 - 6 | 468 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
