Assay Details
| Gene Symbol: AT3G60415 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115906.4 | NP_191601.1 | 1 - 2 | 353 | 66 | |
| GenBank mRNA | AK119045.1 | - | 1 - 2 | 367 | 66 | |
| BT006233.1 | - | 1 - 2 | 331 | 66 | ||
More Information
Set Membership: |
|

AT3G60415
Genomic Map