Assay Details
| Gene Symbol: FUS6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115978.4 | NP_567109.1 | 1 - 2 | 559 | 83 | |
| GenBank mRNA | AF360295.1 | - | 1 - 2 | 445 | 83 | |
| AF395057.1 | - | 1 - 2 | 391 | 83 | ||
| AY051056.1 | - | 1 - 2 | 391 | 83 | ||
More Information
Set Membership: |
|

FUS6
Genomic Map