Assay Details
Gene Symbol: HDG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115979.5 | NP_191674.1 | 2 - 3 | 812 | 72 | |
GenBank mRNA | AY050866.1 | - | 2 - 3 | 533 | 72 | |
AY096757.1 | - | 2 - 3 | 374 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
