Assay Details
| Gene Symbol: HDG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115979.5 | NP_191674.1 | 3 - 4 | 936 | 80 | |
| GenBank mRNA | AY050866.1 | - | 3 - 4 | 657 | 80 | |
| AY096757.1 | - | 3 - 4 | 498 | 80 | ||
More Information
Set Membership: |
|

HDG1
Genomic Map