Assay Details
| Gene Symbol: HDG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_115979.5 | NP_191674.1 | 6 - 7 | 1927 | 72 | |
| GenBank mRNA | AY050866.1 | - | 6 - 7 | 1648 | 72 | |
| AY096757.1 | - | 6 - 7 | 1489 | 72 | ||
More Information
Set Membership: |
|

HDG1
Genomic Map