Assay Details
Gene Symbol: HDG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115979.5 | NP_191674.1 | 7 - 8 | 2101 | 86 | |
GenBank mRNA | AY050866.1 | - | 7 - 8 | 1822 | 86 | |
AY096757.1 | - | 7 - 8 | 1663 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
