Assay Details
Gene Symbol: AT3G61290 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115993.2 | NP_191688.1 | 2 - 3 | 712 | 90 | |
GenBank mRNA | KJ138779.1 | - | 2 - 3 | 712 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
