Assay Details
| Gene Symbol: ZIP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128563.2 | NP_180569.1 | 1 - 2 | 842 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZIP6| Gene Symbol: ZIP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128563.2 | NP_180569.1 | 1 - 2 | 842 | 59 | |
Set Membership: |
|