Assay Details
| Gene Symbol: EMB3147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128573.4 | NP_565697.1 | 1 - 2 | 586 | 70 | |
| NM_201835.2 | NP_973564.2 | 1 - 2 | 578 | 70 | ||
| GenBank mRNA | AY091077.1 | - | 1 - 2 | 342 | 70 | |
| AY122949.1 | - | 1 - 2 | 292 | 70 | ||
| AY735592.1 | - | 1 - 2 | 310 | 70 | ||
| AY924753.1 | - | 1 - 2 | 292 | 70 | ||
More Information
Set Membership: |
|

EMB3147
Genomic Map