Assay Details
| Gene Symbol: FC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202710.1 | NP_001189639.1 | 1 - 2 | 209 | 129 | |
| NM_128592.3 | NP_180598.1 | 1 - 2 | 243 | 129 | ||
| GenBank mRNA | BT000465.1 | - | 1 - 2 | 186 | 129 | |
| BT008877.1 | - | 1 - 2 | 80 | 129 | ||
More Information
Set Membership: |
|

FC2
Genomic Map