Assay Details
| Gene Symbol: FC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_128592.3 | NP_180598.1 | 6 - 7 | 1023 | 79 | |
| GenBank mRNA | BT000465.1 | - | 6 - 7 | 966 | 79 | |
| BT008877.1 | - | 6 - 7 | 860 | 79 | ||
More Information
Set Membership: |
|

FC2
Genomic Map