Assay Details
Gene Symbol: AK-LYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121331.3 | NP_196832.1 | 2 - 3 | 688 | 107 | |
GenBank mRNA | AY057674.1 | - | 2 - 3 | 562 | 107 | |
BT000493.1 | - | 2 - 3 | 484 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
