Assay Details
| Gene Symbol: AK-LYS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121331.3 | NP_196832.1 | 5 - 6 | 1036 | 106 | |
| GenBank mRNA | AY057674.1 | - | 5 - 6 | 910 | 106 | |
| BT000493.1 | - | 5 - 6 | 832 | 106 | ||
More Information
Set Membership: |
|

AK-LYS1
Genomic Map