Assay Details
Gene Symbol: AK-LYS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121331.3 | NP_196832.1 | 9 - 10 | 1462 | 68 | |
GenBank mRNA | AY057674.1 | - | 9 - 10 | 1336 | 68 | |
BT000493.1 | - | 9 - 10 | 1258 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
