Assay Details
| Gene Symbol: CRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001085108.1 | NP_001078577.1 | 1 - 2 | 579 | 106 | |
| NM_121332.4 | NP_196833.2 | 1 - 2 | 1521 | 106 | ||
More Information
Set Membership: |
|

CRN
Genomic Map