Assay Details
| Gene Symbol: CRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121332.4 | NP_196833.2 | 2 - 3 | 1990 | 94 | |
| NM_180481.4 | NP_850812.2 | 1 - 2 | 1927 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CRN| Gene Symbol: CRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121332.4 | NP_196833.2 | 2 - 3 | 1990 | 94 | |
| NM_180481.4 | NP_850812.2 | 1 - 2 | 1927 | 94 | ||
Set Membership: |
|