Assay Details
| Gene Symbol: SFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121333.4 | NP_196834.3 | 13 - 14 | 1780 | 145 | |
| GenBank mRNA | AB194395.1 | - | 13 - 14 | 1230 | 145 | |
More Information
Set Membership: |
|

SFC
Genomic Map