Assay Details
| Gene Symbol: SFC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121333.4 | NP_196834.3 | 14 - 15 | 1867 | 113 | |
| GenBank mRNA | AB194395.1 | - | 14 - 15 | 1317 | 113 | |
More Information
Set Membership: |
|

SFC
Genomic Map