Assay Details
| Gene Symbol: PAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121373.4 | NP_196874.1 | 1 - 2 | 165 | 100 | |
| GenBank mRNA | BT026370.1 | - | 1 - 2 | 44 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PAO1| Gene Symbol: PAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121373.4 | NP_196874.1 | 1 - 2 | 165 | 100 | |
| GenBank mRNA | BT026370.1 | - | 1 - 2 | 44 | 100 | |
Set Membership: |
|