Assay Details
| Gene Symbol: PAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121373.4 | NP_196874.1 | 2 - 3 | 414 | 114 | |
| GenBank mRNA | BT026370.1 | - | 2 - 3 | 293 | 114 | |
More Information
Set Membership: |
|

PAO1
Genomic Map