Assay Details
| Gene Symbol: PAO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_121373.4 | NP_196874.1 | 4 - 5 | 596 | 119 | |
| GenBank mRNA | BT026370.1 | - | 4 - 5 | 475 | 119 | |
More Information
Set Membership: |
|

PAO1
Genomic Map