Assay Details
| Gene Symbol: FZR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001203376.1 | NP_001190305.1 | 7 - 8 | 1524 | 72 | |
| NM_121387.4 | NP_196888.2 | 6 - 7 | 1676 | 72 | ||
| GenBank mRNA | AY099581.1 | - | 6 - 7 | 1676 | 72 | |
| BT002165.1 | - | 6 - 7 | 1391 | 72 | ||
More Information
Set Membership: |
|

FZR3
Genomic Map